DL-Menthol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol
A231109-25G
A231109-1000G
A231109-100G
A231109-500G
77089-374EA
11
USD
77089-374
77165-530
77165-532
77165-534
DL-Menthol 97%
DL-Menthol
Caution: for Research Use Only
Formula:
C₁₀H₂₀O MW: 156.27 g/mol |
MDL Number:
MFCD00001484 CAS Number: 89-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...