2-Amino-3-fluoro-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

Synonyms: 3-Fluoro-5-(trifluoromethyl)pyridin-2-amine

A903778-1G A903778-250MG A903778-25G A903778-5G
77036-852EA 11 USD
77036-852 77036-854 77036-856 77036-858
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine 98%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine

Caution: for Research Use Only

Formula: C₆H₄F₄N₂
MDL Number: MFCD10699117
CAS Number: 852062-17-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR