4-Biphenylmethyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Chloromethyl)biphenyl

A858410-1G A858410-25G A858410-5G
77006-874EA 18 USD
77006-874 77006-876 77006-878
4-Biphenylmethyl chloride 98%
4-Biphenylmethyl chloride

Caution: for Research Use Only

Formula: C₁₃H₁₁Cl
MDL Number: MFCD00000917
CAS Number: 1667-11-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR