2,2,6,6-Tetramethylpiperidin-4-ol 98%

Supplier: Ambeed
Danger

Synonyms: 2,2,6,6-Tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethylpiperidine

A850108-25G A850108-100G
77036-680EA 5.31 USD
77036-680 77552-596 77873-313
2,2,6,6-Tetramethylpiperidin-4-ol 98%
2,2,6,6-Tetramethylpiperidin-4-ol

Caution: for Research Use Only

Formula: C₉H₁₉NO
MW: 157.26 g/mol
MDL Number: MFCD00005983
CAS Number: 2403-88-5
UN: 3259
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR