8-Anilino-1-naphthalenesulfonic acid 95%

Supplier: Ambeed
Warning

Synonyms: 8-Anilinonaphthalene-1-sulfonic acid, 8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulfonic acid

A666405-100G A666405-25G A666405-500G A666405-5G
77035-566EA 47 USD
77035-566 77035-568 77035-570 77035-572
8-Anilino-1-naphthalenesulfonic acid 95%
8-Anilino-1-naphthalenesulphonic acid

Caution: for Research Use Only

Formula: C₁₆H₁₃NO₃S
MDL Number: MFCD00003998
CAS Number: 82-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR