9,10-Dicyanoanthracene 98%

Supplier: Ambeed
Danger

Synonyms: 9,10-Anthracenedicarbonitrile, Anthracene-9,10-dicarbonitrile

A636134-1G A636134-250MG A636134-5G A636134-25G
77035-364EA 39 USD
77035-364 77035-366 77035-368 77873-033
9,10-Dicyanoanthracene 98%
9,10-Dicyanoanthracene

Caution: for Research Use Only

Formula: C₁₆H₈N₂
MW: 228.25 g/mol
MDL Number: MFCD00041733
CAS Number: 1217-45-4
UN: 3439
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR