4-Amino-o-anisic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-methoxybenzenecarboxylic acid

A593183-1G A593183-5G A593183-25G
77005-338EA 17 USD
77005-338 77673-590 77882-406
4-Amino-o-anisic acid 97%
4-Amino-o-anisic acid

Caution: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16
MDL Number: MFCD00114479
CAS Number: 2486-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR