5-Fluoro-3-(trifluoromethyl)benzene-1,2-diamine 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Diamino-5-fluorobenzotrifluoride, 1,2-Diamino-5-fluoro-3-(trifluoromethyl)benzene

A580212-100MG A580212-1G A580212-250MG
76991-910EA 29 USD
76991-910 76991-912 76991-914
5-Fluoro-3-(trifluoromethyl)benzene-1,2-diamine 97%
5-Fluoro-3-(trifluoromethyl)benzene-1,2-diamine

Caution: for Research Use Only

Formula: C₇H₆F₄N₂
MDL Number: MFCD03094268
CAS Number: 179062-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR