2-Piperidinoethanol 98%

Supplier: Ambeed
Warning

A546563-10G A546563-25G A546563-500G
77005-058EA 14 USD
77005-058 77005-060 77005-062
2-Piperidinoethanol 98%
2-Piperidinoethanol

Caution: for Research Use Only

Formula: C₇H₁₅NO
MDL Number: MFCD00006512
CAS Number: 3040-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR