2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture) 97%

Supplier: Ambeed
Warning

Synonyms: 2,4,7,9-Tetramethyl-5-decyne-4,7-diol

A515576-500G A515576-100G
77004-802EA 67 USD
77004-802 77882-265
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture) 97%
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso-mixture)

Caution: for Research Use Only

Formula: C₁₄H₂₆O₂
MDL Number: MFCD00008942
CAS Number: 126-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR