3,5-Di-tert-butylsalicylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Bis-tert-butylsalicylic acid, 3,5-Di-tert-butyl-2-hydroxybenzoic Acid

A508067-25G A508067-5G A508067-100G
77004-744EA 10 USD
77004-744 77572-618 77882-252
3,5-Di-tert-butylsalicylic acid 95%
3,5-Di-tert-butylsalicylic acid

Caution: for Research Use Only

Formula: C₁₅H₂₂O₃
MW: 250.34 g/mol
MDL Number: MFCD00059058
CAS Number: 19715-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR