5-Bromo-3-(trifluoromethyl)picolinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-(trifluoromethyl)-2-pyridinecarboxylic acid

A494004-100MG A494004-1G A494004-250MG A494004-5G
77034-432EA 17 USD
77034-432 77034-434 77034-436 77673-282
5-Bromo-3-(trifluoromethyl)picolinic acid 98%
5-Bromo-3-(trifluoromethyl)picolinic acid

Caution: for Research Use Only

Formula: C₇H₃BrF₃NO₂
MW: 270.003 g/mol
MDL Number: MFCD13185433
CAS Number: 1211580-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR