3-Amino-L-tyrosine 97%

Supplier: Ambeed
Warning

A487955-1G A487955-250MG
77004-614EA 129 USD
77004-614 77004-616
3-Amino-L-tyrosine 97%
3-Amino-L-tyrosine

Caution: for Research Use Only

Formula: C₉H₁₂N₂O₃
MDL Number: MFCD00047831
CAS Number: 300-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR