7-Bromo-2H-1,4-benzothiazin-3(4H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-2H-benzo[b][1,4]thiazin-3(4H)-one

A451799-5G A451799-250MG A451799-1G
76990-746EA 51.99 USD
76990-746 76990-748 76990-744
7-Bromo-2H-1,4-benzothiazin-3(4H)-one 98%
7-Bromo-2H-1,4-benzothiazin-3(4H)-one

Caution: for Research Use Only

Formula: C₈H₆BrNOS
MW: 244.11 g/mol
MDL Number: MFCD02660583
CAS Number: 90814-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR