2-Amino-3-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Danger

A442837-1G A442837-25G A442837-5G
77016-686EA 23.34 USD
77016-686 77016-688 77016-690 77016-692
2-Amino-3-(trifluoromethyl)pyridine 97%
2-Amino-3-(trifluoromethyl)pyridine

Caution: for Research Use Only

Formula: C₆H₅F₃N₂
MDL Number: MFCD04972538
CAS Number: 183610-70-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR