7-Aza-spiro[3.5]nonane hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 7-Azaspiro[3.5]nonane hydrochloride

A436120-1G A436120-250MG
76990-574EA 235 USD
76990-574 77572-352
7-Aza-spiro[3.5]nonane hydrochloride 98%
7-Aza-spiro[3.5]nonane hydrochloride

Caution: for Research Use Only

Formula: C₈H₁₅N·ClH
MW: 161.672 g/mol
MDL Number: MFCD23105809
CAS Number: 1414885-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR