4-Iodophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A432658-1G A432658-250MG A432658-25G A432658-5G
77016-596EA 24 USD
77016-596 77016-598 77016-600 77016-602
4-Iodophenylboronic acid pinacol ester 98%
4-Iodophenylboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₂H₁₆BIO₂
MDL Number: MFCD09953499
CAS Number: 73852-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR