5-Bromo-1,7-dimethyl-1H-indazole 96%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1,7-dimethylindazole

A388792-1G A388792-250MG
77033-532EA 73 USD
77033-532 77033-534
5-Bromo-1,7-dimethyl-1H-indazole 96%
5-Bromo-1,7-dimethyl-1H-indazole

Caution: for Research Use Only

Formula: C₉H₉BrN₂
MDL Number: MFCD11226595
CAS Number: 1092352-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR