O-tert-Butyl-L-serine 97%

Supplier: Ambeed
Warning

Synonyms: H-Ser(tBu)-OH

A363988-1G A363988-5G A363988-10G
77003-416EA 5.31 USD
77003-416 77003-418 77601-612
O-tert-Butyl-L-serine 97%
O-tert-Butyl-L-serine

Caution: for Research Use Only

Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
MDL Number: MFCD00066088
CAS Number: 18822-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR