9-Anthracenylmethyl acrylate ≥98%

Supplier: Ambeed
Warning

Synonyms: Anthracen-9-ylmethyl acrylate, Acrylic Acid 9-Anthrylmethyl Ester, Anthracen-9-ylmethyl Prop-2-enoate, 2-propenoic acid 9-anthracenylmethyl ester, 9-Anthrylmethyl Acrylate

A345093-100MG A345093-250MG A345093-5G
77033-064EA 28 USD
77033-064 77033-066 77033-068
9-Anthracenylmethyl acrylate ≥98%
9-Anthracenylmethyl acrylate

Caution: for Research Use Only

Formula: C₁₈H₁₄O₂
MW: 262.30 g/mol
MDL Number: MFCD04039960
CAS Number: 31645-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR