2-(3-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%

Supplier: Ambeed
Warning

A336295-100MG A336295-1G A336295-250MG
76989-348EA 73 USD
76989-348 76989-350 76989-352
2-(3-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%
2-(3-Aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol

Caution: for Research Use Only

Formula: C₉H₇F₆NO
MDL Number: MFCD17015636
CAS Number: 2402-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR