Risperidone 98%

Supplier: Ambeed
Danger

A324792-10MG A324792-5G A324792-1G
77002-916EA 5 USD
77002-916 77601-456 77672-672
Risperidone 98%
Risperidone

Caution: for Research Use Only

Formula: C₂₃H₂₇FN₄O₂
MDL Number: MFCD00274576
CAS Number: 106266-06-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR