HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
A318293-1000G
A318293-100G
A318293-25G
A318293-500G
77032-718EA
58
USD
77032-718
77032-720
77032-722
77032-724
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid 97%
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)
Caution: for Research Use Only
Formula:
C₈H₁₈N₂O₄S MW: 238.31 g/mol |
MDL Number:
MFCD00006158 CAS Number: 7365-45-9 Merck Index: 13,04672 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...