2-Pyrimidinecarbonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 2-Cyanopyrimidine

A294820-10G A294820-25G A294820-5G A294820-100G
76988-696EA 10 USD
76988-696 76988-698 76988-700 77612-696
2-Pyrimidinecarbonitrile 98%
2-Pyrimidinecarbonitrile

Caution: for Research Use Only

Formula: C₅H₃N₃
MW: 105.1 g/mol
MDL Number: MFCD00160513
CAS Number: 14080-23-0
UN: 3439
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR