IPI-145 98%

Supplier: Ambeed
Warning

Synonyms: (S)-3-(1-((9H-Purin-6-yl)amino)ethyl)-8-chloro-2-phenylisoquinolin-1(2H)-one

A270608-1MG A270608-5MG
77014-414EA 19 USD
77014-414 77014-416
IPI-145 98%
IPI-145

Caution: for Research Use Only

Formula: C₂₂H₁₇ClN₆O
MW: 416.9 g/mol
MDL Number: MFCD15144635
CAS Number: 1201438-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR