2-(4-Aminophenyl)-2-methylpropanenitrile 97%
Supplier: Ambeed
|
Warning
|
A248674-100MG
A248674-1G
A248674-250MG
A248674-5G
77031-460EA
5
USD
77031-460
77031-462
77031-464
77872-239
2-(4-Aminophenyl)-2-methylpropanenitrile 97%
2-(4-Aminophenyl)-2-methylpropanenitrile
Caution: for Research Use Only
Formula:
C₁₀H₁₂N₂ |
MDL Number:
MFCD09909912 CAS Number: 115279-57-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...