2'-Bromo-6'-fluoroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Bromo-6-fluorophenyl)ethanone

A215659-100MG A215659-10G A215659-1G A215659-250MG A215659-5G
77030-662EA 7 USD
77030-662 77030-664 77030-666 77030-668 77030-670
2'-Bromo-6'-fluoroacetophenone 97%
2'-Bromo-6'-fluoroacetophenone

Caution: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD13193629
CAS Number: 928715-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR