AZD1480 98+%

Supplier: Ambeed
Warning

Synonyms: (S)-5-Chloro-N2-(1-(5-fluoropyrimidin-2-yl)ethyl)-N4-(5-methyl-1H-pyrazol-3-yl)pyrimidine-2,4-diamine, AZD-1480

A209618-10MG A209618-1MG A209618-50MG A209618-5MG
76986-678EA 144 USD
76986-678 76986-680 76986-682 76986-684
AZD1480 98+%
AZD1480

Caution: for Research Use Only

Formula: C₁₄H₁₄ClFN₈
MW: 348.77 g/mol
MDL Number: MFCD16038904
CAS Number: 935666-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR