5-Bromo-1,3,4-thiadiazol-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-bromo-1,3,4-thiadiazole, 1,3,4-Thiadiazol-2-amine, 5-bromo-, 5-bromo-1,3,4-thiadiazol-2-amine

A201830-100G A201830-10G A201830-1G A201830-25G A201830-5G
76999-356EA 55 USD
76999-356 76999-358 76999-360 76999-362 76999-364
5-Bromo-1,3,4-thiadiazol-2-amine 98%
5-Bromo-1,3,4-thiadiazol-2-amine

Caution: for Research Use Only

Formula: C₂H₂BrN₃S
MDL Number: MFCD00464325
CAS Number: 37566-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR