4-Amino-5-bromo-2-chloropyrimidine 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A195986-100G A195986-10G A195986-1G A195986-250MG A195986-5G A195986-25G
77012-504EA 267 USD
77012-504 77012-506 77012-508 77012-510 77012-512 77683-534
4-Amino-5-bromo-2-chloropyrimidine 97%, Ambeed.Inc
4-Amino-5-bromo-2-chloropyrimidine

Caution: For research use only.

Formula: C₄H₃BrClN₃
MDL Number: MFCD07782040
CAS Number: 205672-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR