Anacardic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-6-pentadecylbenzoic acid

A190197-10MG A190197-25MG A190197-5MG A190197-1MG
76986-056EA 114 USD
76986-056 76986-058 76986-060 77611-832
Anacardic acid 98%
Anacardic acid

Caution: for Research Use Only

Formula: C₂₂H₃₆O₃
MW: 348.5 g/mol
MDL Number: MFCD07368254
CAS Number: 16611-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR