α-Hydroxy-o-tolyl-β-D-glucopyranoside 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
, 2-(Hydroxymethyl)phenyl-β-D-glucopyranoside
, Salicoside
, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol
, D(-)-Salicin
, D(−)-Salicin
A190057-100G
A190057-1G
A190057-10G
A190057-25G
A190057-1MG
A190057-5G
77012-262EA
6.18
USD
77012-262
77012-260
77012-266
77012-264
77012-268
77012-258
α-Hydroxy-o-tolyl-β-D-glucopyranoside 98%
α-Hydroxy-o-tolyl β-D-glucopyranoside
Caution: for Research Use Only
Formula:
C₁₃H₁₈O₇ MW: 286.28 g/mol |
MDL Number:
MFCD00006590 CAS Number: 138-52-3 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...