Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99%

Supplier: Ambeed
Warning

A186986-100G A186986-25G A186986-500G A186986-5G A186986-1000G
77029-670EA 35 USD
77029-670 77029-672 77029-674 77029-676 77567-728
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99%
Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)

Caution: For research use only.

Formula: C₈H₁₉NO₅
MDL Number: MFCD00002853
CAS Number: 6976-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR