5,5'-Bis(bromomethyl)-2,2'-bipyridine 97%

Supplier: Ambeed
Danger

A186985-100MG A186985-1G A186985-5G
77029-668EA 37.14 USD
77029-668 77600-520 77667-556
5,5'-Bis(bromomethyl)-2,2'-bipyridine 97%
5,5'-Bis(bromomethyl)-2,2'-bipyridine

Caution: for Research Use Only

Formula: C₁₂H₁₀Br₂N₂
MW: 342.03 g/mol
MDL Number: MFCD12031102
CAS Number: 92642-09-6
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR