Polyethylene glycol monomethyl ether, M.W. ∼750
Supplier: Ambeed
|
Warning
|
A186030-10G
A186030-25G
A186030-500G
A186030-1000G
A186030-100G
77012-118EA
6
USD
77012-118
77012-120
77012-122
77567-712
77611-796
Polyethylene glycol monomethyl ether, M.W. ∼750
Polyethylene glycol monomethyl ether
Caution: For research use only.
Formula:
CH₃(OCH₂CH₂)nOH |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...