2-Amino-3-iodopyridine 98%
Supplier: Ambeed
|
Warning
|
A173148-100G
A173148-10G
A173148-1G
A173148-5G
A173148-25G
76998-278EA
247
USD
76998-278
76998-280
76998-282
76998-284
76998-286
77671-460
2-Amino-3-iodopyridine 98%
2-Amino-3-iodopyridine
Caution: for Research Use Only
Formula:
C₅H₅IN₂ |
MDL Number:
MFCD01651885 CAS Number: 104830-06-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...