L-Valine-tert-butyl ester hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Val-OtBu·HCl
, L-2-Aminoisovaleric Acid tert-Butyl Ester Hydrochloride
, (S)-tert-Butyl 2-amino-3-methylbutanoate hydrochloride
A171504-100G
A171504-25G
A171504-1G
A171504-5G
A171504-500G
A171504-10G
76998-228EA
26.78
USD
76998-228
76998-224
76998-226
76998-232
76998-222
76998-230
L-Valine-tert-butyl ester hydrochloride 95%
L-Valine-tert-butyl ester hydrochloride
Caution: for Research Use Only
Formula:
C₉H₁₉NO₂·HCl MW: 209.7 g/mol |
MDL Number:
MFCD00034863 CAS Number: 13518-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...