5-Amino-6-chloroquinoline 98%
Supplier: Ambeed
|
Warning
|
A168523-1G
A168523-250MG
A168523-5G
77028-918EA
27
USD
77028-918
77028-920
77028-922
5-Amino-6-chloroquinoline 98%
5-Amino-6-chloroquinoline
Caution: for Research Use Only
Formula:
C₉H₇ClN₂ MW: 178.6200 g/mol |
MDL Number:
MFCD02947117 CAS Number: 341010-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...