2-Amino-6-bromophenol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-2-hydroxyaniline
A167777-100G
A167777-10G
A167777-1G
A167777-250MG
A167777-25G
A167777-500G
A167777-5G
76998-078EA
228
USD
76998-078
76998-080
76998-082
76998-084
76998-086
76998-088
76998-090
2-Amino-6-bromophenol 97%
2-Amino-6-bromophenol
Caution: for Research Use Only
Formula:
C₆H₆BrNO MW: 188.02 g/mol |
MDL Number:
MFCD08277300 CAS Number: 28165-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...