5-Aza-2'-deoxycytidine 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one
A156645-100MG
A156645-10MG
A156645-1MG
A156645-1G
A156645-250MG
A156645-50MG
A156645-5G
76997-554EA
25
USD
76997-554
76997-556
76997-558
77549-988
77600-238
77600-240
77671-210
5-Aza-2'-deoxycytidine 99%
5-Aza-2'-deoxycytidine
Caution: for Research Use Only
Formula:
C₈H₁₂N₄O₄ |
MDL Number:
MFCD00043011 CAS Number: 2353-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...