Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Lys-OH.HClNɑ-Fmoc-L-lysine HydrochlorideNα-Fmoc-L-lysine hydrochlorideFmoc-Lys-OH·HCl

A146292-5G A146292-100G A146292-1G A146292-10G A146292-25G
76997-094EA 178.24 USD
76997-094 76997-102 76997-100 76997-098 76997-096
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride 98%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride

Caution: for Research Use Only

Formula: C₂₁H₂₄N₂O₄
MW: 404.89 g/mol
MDL Number: MFCD00190889
CAS Number: 139262-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR