Azetidin-3-ylmethanol hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-Azetidinemethanol hydrochloride

A141778-100G A141778-100MG A141778-10G A141778-1G A141778-250MG A141778-25G A141778-5G
77027-590EA 545 USD
77027-590 77027-592 77027-594 77027-596 77027-598 77027-600 77027-602
Azetidin-3-ylmethanol hydrochloride 97%
Azetidin-3-ylmethanol hydrochloride

Caution: for Research Use Only

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD09743446
CAS Number: 928038-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR