(4-Aminopyridin-3-yl)methanol 95%

Supplier: Ambeed
Danger

Synonyms: 4-Amino-3-pyridinemethanol, (4-Amino-pyridin-3-yl)-methanol

A137500-1G A137500-250MG
77010-192EA 81 USD
77010-192 77889-927
(4-Aminopyridin-3-yl)methanol 95%
(4-Aminopyridin-3-yl)methanol

Caution: for Research Use Only

Formula: C₆H₈N₂O
MDL Number: MFCD06203066
CAS Number: 138116-34-4
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR