Fmoc-β-(3-benzothienyl)-Ala-OH 97%

Supplier: Ambeed
Warning

A132473-100MG A132473-1G A132473-250MG A132473-5G
76996-458EA 10 USD
76996-458 76996-460 76996-462 76996-464
Fmoc-β-(3-benzothienyl)-Ala-OH 97%
Fmoc-beta-(3-benzothienyl)-Ala-OH

Caution: for Research Use Only

Formula: C₂₆H₂₁NO₄S
MDL Number: MFCD00672562
CAS Number: 177966-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR