Pyrazinamide 98%

Supplier: Ambeed
Warning

Synonyms: Pyrazinecarboxamide, Pyrazine-2-carboxamide, 2-Pyrazinecarboxamide

A132133-100G A132133-10G A132133-250MG A132133-500G A132133-25G A132133-1000G
77027-092EA 21.22 USD
77027-092 77027-094 77027-096 77027-098 77314-248 77670-866
Pyrazinamide 98%
Pyrazinamide

Caution: for Research Use Only

Formula: C₅H₅N₃O
MW: 123.11 g/mol
MDL Number: MFCD00006132
CAS Number: 98-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR