2-Amino-4,6-dichloropyridine 97%

Supplier: Ambeed
Warning

A129735-100G A129735-10G A129735-1G A129735-250MG A129735-25G A129735-5G
76983-600EA 499 USD
76983-600 76983-602 76983-604 76983-606 76983-608 76983-610
2-Amino-4,6-dichloropyridine 97%
2-Amino-4,6-dichloropyridine

Caution: for Research Use Only

Formula: C₅H₄Cl₂N₂
MDL Number: MFCD04037218
CAS Number: 116632-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR