2-Amino-5-iodo-4-methoxypyrimidine 98%

Supplier: Ambeed
Warning

A129201-1G A129201-250MG A129201-5G
77026-974EA 54 USD
77026-974 77026-976 77026-978
2-Amino-5-iodo-4-methoxypyrimidine 98%
2-Amino-5-iodo-4-methoxypyrimidine

Caution: for Research Use Only

Formula: C₅H₆IN₃O
MDL Number: MFCD11520889
CAS Number: 89322-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR