3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Diethyl (4-oxobenzo[d][1,2,3]triazin-3(4H)-yl) phosphate, DEPBT, Diethyl 3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl Phosphate
A126046-100G
A126046-10G
A126046-25G
A126046-500G
A126046-5G
76996-126EA
102.91
USD
76996-126
76996-128
76996-130
76996-132
76996-134
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one 98%
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one
Caution: for Research Use Only
Formula:
C₁₁H₁₄N₃O₅P MW: 299.22 g/mol |
MDL Number:
MFCD01236967 CAS Number: 165534-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...