tert-Butyl (piperazin-1-yl)acetate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(piperazin-1-yl)acetate, 1-[(tert-Butoxycarbonyl)methyl]piperazine, (Piperazin-1-yl)acetic acid tert-butyl ester

A114032-100MG A114032-1G A114032-250MG A114032-5G A114032-25G
77025-942EA 10 USD
77025-942 77025-944 77025-946 77025-948 77570-766
tert-Butyl (piperazin-1-yl)acetate 95%
tert-Butyl (piperazin-1-yl)acetate

Caution: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD06659535
CAS Number: 112257-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR