4-(alpha-Methylbenzyl)resorcinol 98%

Supplier: Ambeed
Warning

Synonyms: 4-(α-Methylbenzyl)resorcinol, 4-(1-Phenylethyl)resorcinol, 4-(1-Phenylethyl)benzene-1,3-diol, 4-(1-Phenylethyl)resorcino

A108250-100G A108250-10G A108250-1G A108250-250MG A108250-25G A108250-5G
77008-760EA 108 USD
77008-760 77008-762 77008-764 77008-766 77008-768 77008-770
4-(alpha-Methylbenzyl)resorcinol 98%
4-(alpha-Methylbenzyl)resorcinol

Caution: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
MDL Number: MFCD21648466
CAS Number: 85-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR